Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-2-(tert-butyl)benzene 95%

Supplier: AMBEED, INC
Warning

A337764-1G A337764-5G A337764-100MG A337764-250MG
103825-756EA 143 USD InStock
103825-756 103836-728 76839-744 76839-746 77015-506 77015-508
1-Bromo-2-(tert-butyl)benzene 95%
1-Bromo-2-(tert-butyl)benzene
Precaución: for Research Use Only


Formula: C₁₀H₁₃Br
MW: 213.11 g/mol
Boiling Pt: 221.0±9.0 °C (760 mm Hg)
Flash Pt: 86.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00234660
CAS Number: 7073-99-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty