Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2,3-difluoroanisole 97%

Supplier: AMBEED, INC
Warning

A192598-1G A192598-5G A192598-100G A192598-250MG A192598-1G A192598-25G A192598-5G
103816-464EA 8 USD InStock
103816-464 103847-074 77139-110 77139-112 77302-308 77302-310 77302-312
4-Bromo-2,3-difluoroanisole 97%
4-Bromo-2,3-difluoroanisole
Precaución: for Research Use Only


Formula: C₇H₅BrF₂O
MW: 223.02
Boiling Pt: 203.2 °C (760 mm Hg)
Flash Pt: 91.1 °C
Storage Temperature: Ambient
MDL Number: MFCD07368017
CAS Number: 406482-22-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty