Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-fluorotoluene 98%

Supplier: AMBEED, INC
Warning

A152173-5G A152173-10G A152173-1G A152173-25G
103842-512EA 10 USD InStock
103842-512 77137-814 77137-816 77318-122
3-Bromo-2-fluorotoluene 98%
3-Bromo-2-fluorotoluene
Precaución: for Research Use Only


Formula: C₇H₆BrF
MW: 189.02 g/mol
Boiling Pt: 118…123 °C (760 mm Hg)
Flash Pt: 76.7 °C
Storage Temperature: Ambient
MDL Number: MFCD04972694
CAS Number: 59907-12-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty