Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-iodotoluene 98%

Supplier: AMBEED, INC
Warning

A309684-1G A309684-1G A309684-5G A309684-250MG A309684-25G
103814-594EA 49 USD InStock
103814-594 103835-774 76838-542 76838-544 77167-090 77167-092
3-Bromo-2-iodotoluene 98%
3-Bromo-2-iodotoluene
Precaución: for Research Use Only


Formula: C₇H₆BrI
Storage Temperature: Refrigerator
MDL Number: MFCD12547770
CAS Number: 869500-07-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty