Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-3,3-dimethylbutane 98%

Supplier: AMBEED, INC
Danger

A471994-1G A471994-25G A471994-5G A471994-10G
103828-036EA 5 USD InStock
103828-036 103839-112 76843-020 76843-022 76843-024 77241-244
1-Bromo-3,3-dimethylbutane 98%
1-Bromo-3,3-dimethylbutane
Precaución: for Research Use Only


Formula: C₆H₁₃Br
MW: 165.07 g/mol
Boiling Pt: 138 °C (760 mm Hg)
Flash Pt: 33.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00060844
CAS Number: 1647-23-0
UN: 1993
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty