Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-6-chloroanisole 95%

Supplier: AMBEED, INC
Warning

A341979-5G A341979-100MG A341979-250MG A341979-25G A341979-1G
103825-852EA 28 USD InStock
103825-852 103836-826 76828-512 76828-514 76828-516 76839-856
2-Bromo-6-chloroanisole 95%
2-Bromo-6-chloroanisole
Precaución: for Research Use Only


Formula: C₇H₆BrClO
MW: 221.48 g/mol
Boiling Pt: 235.8±20.0 °C (760 mm Hg)
Flash Pt: 96.4 °C
Storage Temperature: Ambient
MDL Number: MFCD15143586
CAS Number: 174913-10-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty