Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-3-ethylbenzene 98%

Supplier: AMBEED, INC
Warning

A487852-1G A487852-5G A487852-1G A487852-25G A487852-10G
103828-190EA 5 USD InStock
103828-190 103839-264 76843-214 76843-216 77241-392
1-Bromo-3-ethylbenzene 98%
1-Bromo-3-ethylbenzene
Precaución: for Research Use Only


Formula: C₈H₉Br
MW: 185.06
Boiling Pt: 204.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00156128
CAS Number: 2725-82-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty