Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-fluoroanisole 96%

Supplier: AMBEED, INC
Warning

A363050-5G A363050-100G A363050-10G A363050-1G A363050-500G A363050-25G
103837-350EA 8 USD InStock
103837-350 76773-358 76773-360 76773-362 76773-364 76840-570
3-Bromo-5-fluoroanisole 96%
3-Bromo-5-fluoroanisole
Precaución: for Research Use Only


Formula: C₇H₆BrFO
MW: 205.024 g/mol
Boiling Pt: 186.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01861129
CAS Number: 29578-39-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty