Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-3-iodobenzene 98% stabilised

Supplier: AMBEED, INC
Warning

Stabilization: Stabilised with copper chip

A147164-5G A147164-1000G A147164-100G A147164-10G A147164-1G A147164-500G A147164-25G A147164-5G
103841-842EA 6 USD InStock
103841-842 77110-080 77110-082 77110-084 77110-086 77110-088 77317-206 77317-208
1-Bromo-3-iodobenzene 98% stabilised
1-Bromo-3-iodobenzene
Precaución: for Research Use Only


Formula: C₆H₄BrI
MW: 282.91 g/mol
Boiling Pt: 240
Melting Pt: -9.3
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00001043
CAS Number: 591-18-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty