Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromophenyl methyl sulphone 98%

Supplier: AMBEED, INC
Warning

A185566-5G A185566-100G A185566-1G A185566-5G
103846-434EA 15 USD InStock
103846-434 76769-718 76769-720 77084-070
4-Bromophenyl methyl sulphone 98%
4-Bromophenyl methyl sulphone
Precaución: for Research Use Only


Formula: C₇H₇BrO₂S
Boiling Pt: 348.6 °C (760 mm Hg)
Melting Pt: 103…107 °C
Flash Pt: 164.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00025065
CAS Number: 3466-32-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty