Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2,3-difluorophenetole 97%

Supplier: AMBEED, INC
Warning

A289239-1G A289239-5G A289239-100G A289239-1G A289239-25G A289239-5G
103813-820EA 5 USD InStock
103813-820 103834-960 77239-010 77305-550 77305-552 77305-554
4-Bromo-2,3-difluorophenetole 97%
4-Bromo-2,3-difluorophenetole
Precaución: for Research Use Only


Formula: C₈H₇BrF₂O
Boiling Pt: 222.7 °C (760 mm Hg)
Flash Pt: 107.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368018
CAS Number: 156573-09-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty