Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-4-fluoro-2-methyl-3-nitrobenzene 97%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A136046-1G A136046-100MG A136046-250MG A136046-5G
103802-264EA 120 USD InStock
103802-264 76983-986 76983-988 77683-201
1-Bromo-4-fluoro-2-methyl-3-nitrobenzene 97%, Ambeed.Inc
1-Bromo-4-fluoro-2-methyl-3-nitrobenzene
Precaución: For research use only.


Formula: C₇H₅BrFNO₂
MW: 234.02 g/mol
Boiling Pt: 269.7±35.0 °C (760 mm Hg)
Flash Pt: 116.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD20486202
CAS Number: 1227210-35-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty