Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-nitroanisole 97%

Supplier: AMBEED, INC
Warning

A179157-5G A179157-25G A179157-100G A179157-10G A179157-500G A179157-5G
103845-746EA 5 USD InStock
103845-746 77083-200 77138-640 77138-642 77138-644 77301-598
4-Bromo-3-nitroanisole 97%
4-Bromo-3-nitroanisole
Precaución: for Research Use Only


Formula: C₇H₆BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD00051511
CAS Number: 5344-78-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty