Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-5-isopropoxy-2-methyl-4-nitrobenzene 97%

Supplier: AMBEED, INC
Warning

A205964-5G A205964-25G A205964-5G
103817-674EA 8 USD InStock
103817-674 103848-346 77302-974 77302-976
1-Bromo-5-isopropoxy-2-methyl-4-nitrobenzene 97%
1-Bromo-5-isopropoxy-2-methyl-4-nitrobenzene
Precaución: for Research Use Only


Formula: C₁₀H₁₂BrNO₃
Boiling Pt: 342.1±37.0 °C (760 mm Hg)
Flash Pt: 160.7 °C
Storage Temperature: Ambient
MDL Number: MFCD28405053
CAS Number: 1202858-68-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty