Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-1,3-difluorobenzene 97%

Supplier: AMBEED, INC
Danger

A590388-5G A590388-25G A590388-10G
103840-068EA 7 USD InStock
103840-068 76844-218 77686-499
5-Chloro-1,3-difluorobenzene 97%
5-Chloro-1,3-difluorobenzene
Precaución: for Research Use Only


Formula: C₆H₃ClF₂
Boiling Pt: 111…112 °C (760 mm Hg)
Flash Pt: 29.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00041518
CAS Number: 1435-43-4
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty