Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Chloro-9H-carbazole 97%

Supplier: AMBEED, INC
Warning

A167766-1G A167766-1G A167766-100MG A167766-250MG
103806-218EA 124 USD InStock
103806-218 77081-512 77247-892 77247-894
1-Chloro-9H-carbazole 97%
1-Chloro-9H-carbazole
Precaución: for Research Use Only


Formula: C₁₄H₂₀O₁₀
Boiling Pt: 388.7±15.0 °C (760 mm Hg)
Melting Pt: 125 °C
Flash Pt: 221 °C
Storage Temperature: Ambient
MDL Number: MFCD18450171
CAS Number: 5599-70-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty