Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Cyano-4-(dimethylamino)pyridiniumtetrafluoroborate 95%

Supplier: AMBEED, INC
Danger

A459355-1G A459355-1G A459355-250MG A459355-5G
103827-900EA 67 USD InStock
103827-900 77307-422 77626-916 77880-001
1-Cyano-4-(dimethylamino)pyridiniumtetrafluoroborate 95%
1-Cyano-4-(dimethylamino)pyridiniumtetrafluoroborate
Precaución: for Research Use Only


Formula: C₈H₁₀BF₄N₃
Melting Pt: 196…200 °C
Storage Temperature: Freezer
MDL Number: MFCD00011998
CAS Number: 59016-56-7
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty