Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Cyclopropyl-6,7-difluoro-8-methoxy-4-oxo-1,4-dihydroquinoline-3-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A491721-25G A491721-5G A491721-100G A491721-10G
103839-294EA 7 USD InStock
103839-294 76843-256 76843-258 77143-954 77143-956
1-Cyclopropyl-6,7-difluoro-8-methoxy-4-oxo-1,4-dihydroquinoline-3-carboxylic acid 98%
1-Cyclopropyl-6,7-difluoro-8-methoxy-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₄H₁₁F₂NO₄
Boiling Pt: 245.2 °C (760 mm Hg)
Melting Pt: 192…194 °C
Flash Pt: 102.1 °C
Storage Temperature: Ambient
MDL Number: MFCD05864420
CAS Number: 112811-72-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty