Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Dodecylimidazole 97%

Supplier: AMBEED, INC
Warning

A132963-1G A132963-10G A132963-5G
103801-806EA 20 USD InStock
103801-806 77234-586 77314-392
1-Dodecylimidazole 97%
1-Dodecylimidazole
Precaución: for Research Use Only


Formula: C₁₅H₂₈N₂
Boiling Pt: 146 °C (0.5 mm Hg)
Flash Pt: 174.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00278859
CAS Number: 4303-67-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty