Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Iodophenetole 98%

Supplier: AMBEED, INC
Warning

A108104-1000G A108104-5G A108104-100G A108104-10G A108104-25G
103807-990EA 462 USD InStock
103807-990 103823-072 77283-686 77283-688 77283-690
4-Iodophenetole 98%
4-Iodophenetole
Precaución: for Research Use Only


Formula: C₈H₉IO
Boiling Pt: 251 °C (760 mm Hg)
Melting Pt: 25…28 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00061124
CAS Number: 699-08-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty