Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Ethyl-1,3-dihydro-2H-benzimidazol-2-one 97%

Supplier: AMBEED, INC
Warning

A147983-1G A147983-5G A147983-250MG A147983-1G A147983-25G A147983-5G
103803-858EA 31 USD InStock
103803-858 103841-974 77235-286 77317-398 77317-400 77317-402
1-Ethyl-1,3-dihydro-2H-benzimidazol-2-one 97%
1-Ethyl-1,3-dihydro-2H-benzimidazol-2-one
Precaución: for Research Use Only


Formula: C₉H₁₀N₂O
Melting Pt: 118…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00005715
CAS Number: 10045-45-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty