Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethylanisole 98%

Supplier: AMBEED, INC
Warning

A855524-25G A855524-5G A855524-10G A855524-1G
103841-250EA 7 USD InStock
103841-250 76845-506 76845-508 77294-012 77294-014
4-Ethylanisole 98%
4-Ethylanisole
Precaución: for Research Use Only


Formula: C₉H₁₂O
Boiling Pt: 195.5 °C (760 mm Hg)
Flash Pt: 68.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00015176
CAS Number: 1515-95-3
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty