Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Ethynyl-3,5-difluorobenzene 97%

Supplier: AMBEED, INC
Danger

A148492-1G A148492-5G A148492-10G A148492-250MG A148492-1G A148492-5G
103803-936EA 20 USD InStock
103803-936 103842-048 76783-258 76783-260 76783-262 77299-662 77299-664
1-Ethynyl-3,5-difluorobenzene 97%
1-Ethynyl-3,5-difluorobenzene
Precaución: for Research Use Only


Formula: C₈H₄F₂
Boiling Pt: 123…124 °C (760 mm Hg)
Flash Pt: 33 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04039982
CAS Number: 151361-87-4
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty