Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Ethynyl-4-(methylsulfonyl)benzene 95%

Supplier: AMBEED, INC
Warning

A282439-1G A282439-5G A282439-1G A282439-5G A282439-250MG
103813-592EA 100 USD InStock
103813-592 103853-544 76837-240 76837-242 77032-182
1-Ethynyl-4-(methylsulfonyl)benzene 95%
1-Ethynyl-4-(methylsulfonyl)benzene
Precaución: for Research Use Only


Formula: C₉H₈O₂S
Melting Pt: 95…106 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08703578
CAS Number: 340771-31-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty