Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Fluoro-2-iodo-3-methylbenzene 97%

Supplier: AMBEED, INC
Warning

A121972-1G A121972-10G A121972-1G
103800-162EA 12 USD InStock
103800-162 77136-542 77312-294
1-Fluoro-2-iodo-3-methylbenzene 97%
1-Fluoro-2-iodo-3-methylbenzene
Precaución: for Research Use Only


Formula: C₇H₆FI
MW: 236.0300 g/mol
Boiling Pt: 212 °C (760 mm Hg)
Flash Pt: 85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04973438
CAS Number: 883502-14-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty