Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Fluoro-3-(methylsulfonyl)benzene 95%

Supplier: AMBEED, INC
Warning

A314268-1G A314268-250MG A314268-5G A314268-1G
103825-028EA 95 USD InStock
103825-028 103835-948 76828-062 76838-756 77306-110
1-Fluoro-3-(methylsulfonyl)benzene 95%
1-Fluoro-3-(methylsulfonyl)benzene
Precaución: for Research Use Only


Formula: C₇H₇FO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD11519068
CAS Number: 657-46-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty