Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Fluoro-4-nitronaphthalene 95% (by HPLC)

Supplier: AMBEED, INC
Warning

A323355-5G A323355-1G A323355-100MG A323355-250MG
103825-332EA 36 USD InStock
103825-332 103836-268 76839-188 77239-626 77239-628
1-Fluoro-4-nitronaphthalene 95% (by HPLC)
1-Fluoro-4-nitronaphthalene
Precaución: for Research Use Only


Formula: C₁₀H₆FNO₂
Storage Temperature: Ambient
MDL Number: MFCD00156398
CAS Number: 341-92-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty