Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Fmoc-azetidine-3-carboxylic acid 95%

Supplier: AMBEED, INC
Warning

A215142-1G A215142-5G A215142-250MG A215142-1G A215142-100G
103818-462EA 25 USD InStock
103818-462 103849-166 77030-634 77087-690 77671-920
1-Fmoc-azetidine-3-carboxylic acid 95%
1-Fmoc-azetidine-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₉H₁₇NO₄
Boiling Pt: 543.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01321018
CAS Number: 193693-64-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty