Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α,α,α-Trifluoro-4-tolualdehyde 95%

Supplier: AMBEED, INC
Warning

A226445-5G A226445-100G A226445-10G A226445-25G A226445-5G A226445-500G
103850-152EA 5 USD InStock
103850-152 76975-972 76975-974 77088-880 77088-882 77881-578
α,α,α-Trifluoro-4-tolualdehyde 95%
α,α,α-Trifluoro-4-tolualdehyde
Precaución: for Research Use Only


Formula: C₈H₅F₃O
MW: 174.12 g/mol
Boiling Pt: 176
Melting Pt: 1…2 °C
Flash Pt: 65.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006952
CAS Number: 455-19-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty