Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-1,2,4-Triazole-1-carboxamidine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A421899-5G A421899-25G A421899-100G A421899-10G
103838-460EA 5 USD InStock
103838-460 76842-078 77240-830 77240-832
1H-1,2,4-Triazole-1-carboxamidine hydrochloride 97%
1H-1,2,4-Triazole-1-carboxamidine hydrochloride
Precaución: for Research Use Only


Formula: C₃H₆ClN₅
Boiling Pt: 294 °C (760 mm Hg)
Melting Pt: 215…220 °C
Flash Pt: 131.6 °C
Storage Temperature: Ambient
MDL Number: MFCD03095468
CAS Number: 19503-26-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty