Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Imidazole 99%

Supplier: AMBEED, INC
Danger

A277098-100G A277098-25G
103809-920EA 21 USD InStock
103809-920 77113-978 77305-252
Imidazole 99%
Imidazole
Precaución: for Research Use Only


Formula: C₃H₄N₂
Boiling Pt: 257 °C (760 mm Hg)
Melting Pt: 88…91 °C
Flash Pt: 145 °C
Storage Temperature: Ambient
MDL Number: MFCD00005183
CAS Number: 288-32-4
UN: 3263
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty