Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Indole-3-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A188858-25G A188858-100G A188858-100MG A188858-10G A188858-500G
103846-730EA 6 USD InStock
103846-730 77084-482 77274-368 77274-370 77274-372 77274-374
Indole-3-carboxylic acid 98%
Indole-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₉H₇NO₂
MW: 161.16 g/mol
Boiling Pt: 419.6 °C (760 mm Hg)
Melting Pt: 232…240 °C
Flash Pt: 207.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00005624
CAS Number: 771-50-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty