Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Indole-4-carbaldehyd 97%

Supplier: AMBEED, INC
Warning

A215907-1G A215907-100G A215907-10G A215907-1G A215907-5G A215907-25G
103818-526EA 7 USD InStock
103818-526 103849-236 76691-616 76691-618 77087-798 77087-800 77303-420
Indole-4-carbaldehyd 97%
Indole-4-carbaldehyd
Precaución: for Research Use Only


Formula: C₉H₇NO
MW: 145.16
Melting Pt: 139…143 °C
Storage Temperature: Ambient
MDL Number: MFCD01632221
CAS Number: 1074-86-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty