Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Indole-6-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A120495-1G A120495-100G A120495-25G A120495-500G A120495-5G A120495-1G
103799-944EA 5 USD InStock
103799-944 77271-462 77271-464 77271-466 77297-768 77312-050
Indole-6-carboxylic acid 98%
Indole-6-carboxylic acid
Precaución: for Research Use Only


Formula: C₉H₇NO₂
MW: 161.16
Boiling Pt: 419.6 °C (760 mm Hg)
Melting Pt: 248…254 °C
Flash Pt: 207.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00210441
CAS Number: 1670-82-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty