Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Pyrazol-3(2H)-one 98%

Supplier: AMBEED, INC
Warning

A124366-1G A124366-100G A124366-10G A124366-25G A124366-1G A124366-5G
103800-522EA 6 USD InStock
103800-522 77271-718 77271-720 77271-722 77312-658 77312-660
1H-Pyrazol-3(2H)-one 98%
1H-Pyrazol-3(2H)-one
Precaución: for Research Use Only


Formula: C₃H₄N₂O
Boiling Pt: 326.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00629300
CAS Number: 137-45-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty