Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Pyrazole 98%

Supplier: AMBEED, INC
Warning

A118769-1000G A118769-100G A118769-25G A118769-500G A118769-1000G
103808-190EA 51 USD InStock
103808-190 77094-378 77094-380 77094-382 77311-776
1H-Pyrazole 98%
1H-Pyrazole
Precaución: for Research Use Only


Formula: C₃H₄N₂
MW: 68.08 g/mol
Boiling Pt: 186
Melting Pt: 67…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00005234
CAS Number: 288-13-1
Merck Index: 12,08144
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty