Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Pyrazolo[4,3-b]pyridine-5-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A149159-1G A149159-100MG A149159-250MG A149159-1G
103804-006EA 548 USD InStock
103804-006 76871-524 76871-526 77299-710
1H-Pyrazolo[4,3-b]pyridine-5-carboxylic acid 97%
1H-Pyrazolo[4,3-b]pyridine-5-carboxylic acid
Precaución: for Research Use Only


Formula: C₇H₅N₃O₂
Storage Temperature: Freezer
MDL Number: MFCD18250817
CAS Number: 1260670-03-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty