Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Pyrazolo[4,3-d]pyrimidin-7-amine 97%

Supplier: AMBEED, INC
Danger

A291408-1G A291408-1G A291408-100MG A291408-250MG
103813-916EA 365 USD InStock
103813-916 103835-056 76837-652 76988-670 76988-672
1H-Pyrazolo[4,3-d]pyrimidin-7-amine 97%
1H-Pyrazolo[4,3-d]pyrimidin-7-amine
Precaución: for Research Use Only


Formula: C₅H₅N₅
Boiling Pt: 431.1 °C (760 mm Hg)
Flash Pt: 244.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18809834
CAS Number: 13877-56-0
UN: 3259
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty