Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Pyrrolo[2,3-c]pyridine-5-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A134508-1G A134508-100MG A134508-250MG A134508-1G
103802-018EA 320 USD InStock
103802-018 76823-156 76823-158 77314-778
1H-Pyrrolo[2,3-c]pyridine-5-carboxylic acid 98%
1H-Pyrrolo[2,3-c]pyridine-5-carboxylic acid
Precaución: for Research Use Only


Formula: C₈H₆N₂O₂
MW: 162.14 g/mol
Boiling Pt: 471.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD10697726
CAS Number: 130473-27-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty