Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Pyrrolo[3,2-c]pyridin-4-amine 97%

Supplier: AMBEED, INC
Warning

A129508-1G A129508-100MG A129508-250MG
103801-290EA 128 USD InStock
103801-290 77285-252 77747-925
1H-Pyrrolo[3,2-c]pyridin-4-amine 97%
1H-Pyrrolo[3,2-c]pyridin-4-amine
Precaución: for Research Use Only


Formula: C₇H₇N₃
Boiling Pt: 388.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08690132
CAS Number: 60290-23-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty