Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Pyrrolo[3,4-c]pyridin-3(2H)-one 97%

Supplier: AMBEED, INC
Warning

A149161-1G A149161-5G A149161-100MG A149161-250MG
103804-008EA 51 USD InStock
103804-008 103842-130 77028-042 77028-044
1H-Pyrrolo[3,4-c]pyridin-3(2H)-one 97%
1H-Pyrrolo[3,4-c]pyridin-3(2H)-one
Precaución: for Research Use Only


Formula: C₇H₆N₂O
Boiling Pt: 407.0±34.0 °C (760 mm Hg)
Flash Pt: 200 °C
Storage Temperature: Ambient
MDL Number: MFCD12923778
CAS Number: 40107-95-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty