Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Indancarboxylic acid 98%

Supplier: AMBEED, INC
Warning

A212118-5G A212118-1G A212118-25G A212118-5G A212118-100MG A212118-10G
103818-166EA 21 USD InStock
103818-166 103848-866 77087-314 77087-316 77087-318 77237-408 77237-410
1-Indancarboxylic acid 98%
1-Indancarboxylic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₀O₂
Boiling Pt: 324.6 °C (760 mm Hg)
Flash Pt: 147.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00021239
CAS Number: 14381-42-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty