Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Isopropyl-1H-pyrazol-4-amine 97%

Supplier: AMBEED, INC
Warning

A274742-1G A274742-5G A274742-1G A274742-5G A274742-10G A274742-250MG
103813-214EA 59 USD InStock
103813-214 103853-142 77092-970 77092-972 77289-494 77289-496
1-Isopropyl-1H-pyrazol-4-amine 97%
1-Isopropyl-1H-pyrazol-4-amine
Precaución: for Research Use Only


Formula: C₆H₁₁N₃
Boiling Pt: 234.2 °C (760 mm Hg)
Flash Pt: 95.427 °C
Storage Temperature: Ambient
MDL Number: MFCD08700622
CAS Number: 97421-16-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty