Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxystyrene 98% stabilized

Supplier: AMBEED, INC
Warning

A116963-1G A116963-100G A116963-10G A116963-25G A116963-1G A116963-5G
103798-806EA 6 USD InStock
103798-806 103824-678 77284-580 77284-582 77284-584 77311-466 77311-468
4-Methoxystyrene 98% stabilized
4-Methoxystyrene
Precaución: for Research Use Only


Formula: C₉H₁₀O
Storage Temperature: Refrigerator
MDL Number: MFCD00008619
CAS Number: 637-69-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty