Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-1,2,3,4-tetrahydroquinoxaline dihydrochloride 95%

Supplier: AMBEED, INC
Warning

A140771-1G A140771-1G A140771-5G
103802-902EA 125 USD InStock
103802-902 77316-016 77316-018
1-Methyl-1,2,3,4-tetrahydroquinoxaline dihydrochloride 95%
1-Methyl-1,2,3,4-tetrahydroquinoxaline dihydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₄Cl₂N₂
Melting Pt: 40.5…42 °C
Storage Temperature: Freezer
MDL Number: MFCD08544478
CAS Number: 1259952-24-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty