Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-4,5,6,7-tetrahydro-1H-pyrazolo[3,4-c]pyridine dihydrochloride 95%

Supplier: AMBEED, INC
Warning

A107217-1G A107217-100MG A107217-250MG
103797-152EA 700 USD InStock
103797-152 103822-918 76982-488 76982-490
1-Methyl-4,5,6,7-tetrahydro-1H-pyrazolo[3,4-c]pyridine dihydrochloride 95%
1-Methyl-4,5,6,7-tetrahydro-1H-pyrazolo[3,4-c]pyridine dihydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₃Cl₂N₃
Storage Temperature: Ambient
MDL Number: MFCD09028172
CAS Number: 1228878-69-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty