Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester 95%

Supplier: AMBEED, INC
Warning

A172489-1G A172489-5G A172489-100G A172489-250MG A172489-1G A172489-10G
103806-724EA 25 USD InStock
103806-724 103845-008 76974-074 76974-076 77082-200 77671-450
4-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester 95%
4-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₈H₂₉BN₂O₂
MW: 316.2461 g/mol
Melting Pt: 61…65 °C
Storage Temperature: Ambient
MDL Number: MFCD09266182
CAS Number: 938043-30-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty