Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-5-nitroimidazole 98%

Supplier: AMBEED, INC
Warning

A103808-1G A103808-5G A103808-25G A103808-1G
103796-614EA 9 USD InStock
103796-614 103822-324 77233-164 77309-190
1-Methyl-5-nitroimidazole 98%
1-Methyl-5-nitroimidazole
Precaución: for Research Use Only


Formula: C₄H₅N₃O₂
MW: 127.1000 g/mol
Boiling Pt: 328.9 °C (760 mm Hg)
Melting Pt: 54…57 °C
Flash Pt: 152.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00126968
CAS Number: 3034-42-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty