Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methylimidazole-5-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A311014-1G A311014-5G A311014-1G A311014-25G A311014-5G A311014-100G A311014-10G A311014-500G
103814-648EA 9 USD InStock
103814-648 103835-834 76838-618 76838-620 76838-622 77239-434 77239-436 77239-438
1-Methylimidazole-5-carboxylic acid 97%
1-Methylimidazole-5-carboxylic acid
Precaución: for Research Use Only


Formula: C₅H₆N₂O₂
Boiling Pt: 399 °C (760 mm Hg)
Melting Pt: 247 °C
Flash Pt: 195.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00955677
CAS Number: 41806-40-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty