Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-1-methylpiperidine 98%

Supplier: AMBEED, INC
Warning

A156333-100G A156333-500G A156333-1000G A156333-25G
103808-832EA 135 USD InStock
103808-832 76997-542 76997-544 77318-822 77318-824
4-Hydroxy-1-methylpiperidine 98%
4-Hydroxy-1-methylpiperidine
Precaución: for Research Use Only


Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 200
Storage Temperature: Refrigerator
MDL Number: MFCD00006500
CAS Number: 106-52-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty